Assay Details
Gene Symbol: LOC9271868 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015772830.2 | XP_015628316.1 | 5 - 6 | 741 | 139 | |
GenBank mRNA | AK106686.1 | - | 4 - 5 | 1673 | 139 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
