Assay Details
| Gene Symbol: LOC4334640 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015772633.2 | XP_015628119.1 | 2 - 3 | 593 | 77 | |
| GenBank mRNA | AB022602.1 | - | 2 - 3 | 585 | 77 | |
| AK072053.1 | - | 2 - 3 | 599 | 77 | ||
More Information
Set Membership: |
|

LOC4334640
Genomic Map