Assay Details
| Gene Symbol: LOC4334640 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015772633.2 | XP_015628119.1 | 8 - 9 | 1512 | 78 | |
| GenBank mRNA | AB022602.1 | - | 8 - 9 | 1504 | 78 | |
| AK072053.1 | - | 8 - 9 | 1516 | 78 | ||
More Information
Set Membership: |
|

LOC4334640
Genomic Map