Assay Details
| Gene Symbol: LOC4334647 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015774926.2 | XP_015630412.1 | 2 - 3 | 557 | 93 | |
| GenBank mRNA | AK066364.1 | - | 2 - 3 | 559 | 93 | |
| AK067674.1 | - | 3 - 4 | 389 | 93 | ||
More Information
Set Membership: |
|

LOC4334647
Genomic Map