Assay Details
| Gene Symbol: LOC4334782 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015772693.2 | XP_015628179.1 | 3 - 4 | 739 | 82 | |
| XM_015772694.2 | XP_015628180.1 | 3 - 4 | 707 | 82 | ||
| GenBank mRNA | AK065662.1 | - | 2 - 3 | 696 | 82 | |
More Information
Set Membership: |
|

LOC4334782
Genomic Map