Assay Details
| Gene Symbol: LOC4335111 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015778965.2 | XP_015634451.1 | 4 - 5 | 1503 | 68 | |
| GenBank mRNA | AK119406.1 | - | 4 - 5 | 1490 | 68 | |
More Information
Set Membership: |
|

LOC4335111
Genomic Map