Assay Details
| Gene Symbol: LOC4335342 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015781016.2 | XP_015636502.1 | 13 - 14 | 1215 | 103 | |
| XM_015781018.2 | XP_015636504.1 | 13 - 14 | 1114 | 103 | ||
| XM_015781019.2 | XP_015636505.1 | 11 - 12 | 900 | 103 | ||
| XM_015781021.2 | XP_015636507.1 | 12 - 13 | 980 | 103 | ||
| GenBank mRNA | AK060337.1 | - | 7 - 8 | 455 | 103 | |
| AK069000.1 | - | 13 - 14 | 1146 | 103 | ||
More Information
Set Membership: |
|

LOC4335342
Genomic Map