Assay Details
| Gene Symbol: LOC4335601 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015778796.2 | 5 - 6 | 587 | 113 | ||
| XM_015778797.2 | 5 - 6 | 599 | 113 | |||
| XM_015778798.2 | XP_015634284.1 | 5 - 6 | 586 | 113 | ||
| XM_015778800.2 | XP_015634286.1 | 5 - 6 | 619 | 113 | ||
| XM_015778801.2 | XP_015634287.1 | 5 - 6 | 512 | 113 | ||
More Information
Set Membership: |
|

LOC4335601
Genomic Map