Assay Details
| Gene Symbol: LOC4335611 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015778074.2 | XP_015633560.1 | 11 - 12 | 1139 | 74 | |
| GenBank mRNA | AK061349.1 | - | 10 - 11 | 940 | 74 | |
| AK103713.1 | - | 11 - 12 | 1073 | 74 | ||
More Information
Set Membership: |
|

LOC4335611
Genomic Map