Assay Details
| Gene Symbol: LOC4335611 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015778074.2 | XP_015633560.1 | 8 - 9 | 814 | 71 | |
| GenBank mRNA | AK061349.1 | - | 7 - 8 | 615 | 71 | |
| AK103713.1 | - | 8 - 9 | 748 | 71 | ||
More Information
Set Membership: |
|

LOC4335611
Genomic Map