Assay Details
| Gene Symbol: LOC4335672 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015779138.2 | XP_015634624.1 | 1 | 2037 | 84 | |
| GenBank mRNA | AK109628.1 | - | 1 | 2043 | 84 | |
More Information
Set Membership: |
|

LOC4335672
Genomic Map