Assay Details
| Gene Symbol: LOC4335790 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015778048.2 | XP_015633534.1 | 5 - 6 | 1562 | 69 | |
| GenBank mRNA | AK072276.1 | - | 5 - 6 | 1569 | 69 | |
| AY578159.1 | - | 5 - 6 | 1562 | 69 | ||
| EU095553.1 | - | 5 - 6 | 1517 | 69 | ||
More Information
Set Membership: |
|

LOC4335790
Genomic Map