Assay Details
| Gene Symbol: LOC4335906 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015780146.2 | XP_015635632.1 | 2 - 3 | 777 | 57 | |
| GenBank mRNA | AK101074.1 | - | 2 - 3 | 244 | 57 | |
| AY739308.1 | - | 2 - 3 | 244 | 57 | ||
More Information
Set Membership: |
|

LOC4335906
Genomic Map