Assay Details
| Gene Symbol: LOC9267394 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015781756.2 | XP_015637242.1 | 2 - 3 | 511 | 55 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC9267394| Gene Symbol: LOC9267394 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015781756.2 | XP_015637242.1 | 2 - 3 | 511 | 55 | |
Set Membership: |
|