Assay Details
| Gene Symbol: LOC4336294 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015780084.2 | XP_015635570.1 | 2 - 3 | 393 | 70 | |
| GenBank mRNA | AK067166.1 | - | 2 - 3 | 385 | 70 | |
| AK072162.1 | - | 2 - 3 | 385 | 70 | ||
| AK098918.1 | - | 2 - 3 | 385 | 70 | ||
| AK100589.1 | - | 2 - 3 | 385 | 70 | ||
| JN944362.1 | - | 2 - 3 | 352 | 70 | ||
| KY038503.1 | - | 2 - 3 | 370 | 70 | ||
| Y07766.2 | - | 2 - 3 | 341 | 70 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015780084.2, Y07766.2 transcripts.
Set Membership: |
|

LOC4336294
Genomic Map