Assay Details
Gene Symbol: LOC4336715 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015778882.2 | XP_015634368.1 | 4 - 5 | 675 | 90 | |
XM_015778883.2 | XP_015634369.1 | 4 - 5 | 786 | 90 | ||
GenBank mRNA | AK071401.1 | - | 4 - 5 | 598 | 90 |

More Information
Set Membership: |
![]() ![]() ![]() |
