Assay Details
| Gene Symbol: LOC4336715 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015778882.2 | XP_015634368.1 | 5 - 6 | 792 | 102 | |
| XM_015778883.2 | XP_015634369.1 | 5 - 6 | 903 | 102 | ||
| GenBank mRNA | AK071401.1 | - | 5 - 6 | 715 | 102 | |
More Information
Set Membership: |
|

LOC4336715
Genomic Map