Assay Details
| Gene Symbol: LOC4336715 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015778882.2 | XP_015634368.1 | 6 - 7 | 903 | 76 | |
| XM_015778883.2 | XP_015634369.1 | 6 - 7 | 1014 | 76 | ||
| GenBank mRNA | AK071401.1 | - | 6 - 7 | 826 | 76 | |
More Information
Set Membership: |
|

LOC4336715
Genomic Map