Assay Details
| Gene Symbol: LOC4336726 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK110251.1 | - | 4 | 980 | 137 | |
| Gene Symbol: LOC4336727 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015780022.2 | XP_015635508.1 | 1 | 3161 | 137 | |
More Information
Set Membership: |
|

LOC4336726
Genomic Map