Assay Details
| Gene Symbol: LOC4336744 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015780047.2 | XP_015635533.1 | 3 - 4 | 1646 | 91 | |
| XM_015780048.2 | XP_015635534.1 | 4 - 5 | 808 | 91 | ||
| XM_015780049.2 | XP_015635535.1 | 4 - 5 | 839 | 91 | ||
| GenBank mRNA | AK120597.1 | - | 4 - 5 | 804 | 91 | |
More Information
Set Membership: |
|

LOC4336744
Genomic Map