Assay Details
| Gene Symbol: LOC4337136 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015780184.2 | XP_015635670.1 | 1 - 2 | 698 | 79 | |
| GenBank mRNA | AK060110.1 | - | 1 - 2 | 673 | 79 | |
| AK099507.1 | - | 1 - 2 | 674 | 79 | ||
| AK104741.1 | - | 1 - 2 | 673 | 79 | ||
More Information
Set Membership: |
|

LOC4337136
Genomic Map