Assay Details
| Gene Symbol: LOC4337224 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015780156.2 | XP_015635642.1 | 2 - 3 | 351 | 87 | |
| GenBank mRNA | AK072573.1 | - | 2 - 3 | 358 | 87 | |
| AK109786.1 | - | 2 - 3 | 357 | 87 | ||
More Information
Set Membership: |
|

LOC4337224
Genomic Map