Assay Details
| Gene Symbol: LOC4337765 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015785229.2 | XP_015640715.1 | 4 - 5 | 1150 | 71 | |
| XM_015785230.2 | XP_015640716.1 | 4 - 5 | 1198 | 71 | ||
| XM_015785231.2 | XP_015640717.1 | 4 - 5 | 1193 | 71 | ||
| XM_015785232.2 | XP_015640718.1 | 4 - 5 | 1284 | 71 | ||
| XM_015785233.2 | XP_015640719.1 | 4 - 5 | 1236 | 71 | ||
| XM_015785234.2 | XP_015640720.1 | 3 - 4 | 1101 | 71 | ||
| XM_015785236.2 | XP_015640722.1 | 3 - 4 | 1053 | 71 | ||
| GenBank mRNA | AK065644.1 | - | 1 - 2 | 544 | 71 | |
More Information
Set Membership: |
|

LOC4337765
Genomic Map