Assay Details
Gene Symbol: LOC4337810 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015782944.2 | XP_015638430.2 | 11 - 12 | 850 | 105 | |
GenBank mRNA | AK287699.1 | - | 9 - 10 | 2185 | 105 |

More Information
Set Membership: |
![]() ![]() ![]() |
