Assay Details
Gene Symbol: LOC4337810 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015782944.2 | XP_015638430.2 | 5 - 6 | 416 | 79 | |
GenBank mRNA | AK287699.1 | - | 4 - 5 | 1189 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
