Assay Details
Gene Symbol: LOC4337810 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015782944.2 | XP_015638430.2 | 7 - 8 | 494 | 149 | |
GenBank mRNA | AK287699.1 | - | 6 - 7 | 1267 | 149 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
