Assay Details
| Gene Symbol: LOC4337855 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015782038.2 | XP_015637524.1 | 11 - 12 | 1225 | 118 | |
| XM_015782039.2 | XP_015637525.1 | 9 - 10 | 1065 | 118 | ||
| GenBank mRNA | AK066581.1 | - | 11 - 12 | 1197 | 118 | |
More Information
Set Membership: |
|

LOC4337855
Genomic Map