Assay Details
Gene Symbol: LOC4337855 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015782038.2 | XP_015637524.1 | 3 - 4 | 460 | 111 | |
GenBank mRNA | AK066581.1 | - | 3 - 4 | 432 | 111 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
