Assay Details
Gene Symbol: LOC4337855 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015782038.2 | XP_015637524.1 | 4 - 5 | 538 | 131 | |
GenBank mRNA | AK066581.1 | - | 4 - 5 | 510 | 131 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
