Assay Details
| Gene Symbol: LOC4337855 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015782038.2 | XP_015637524.1 | 8 - 9 | 992 | 74 | |
| XM_015782039.2 | XP_015637525.1 | 6 - 7 | 832 | 74 | ||
| GenBank mRNA | AK066581.1 | - | 8 - 9 | 964 | 74 | |
More Information
Set Membership: |
|

LOC4337855
Genomic Map