Assay Details
Gene Symbol: LOC9266168 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015784463.2 | XP_015639949.1 | 1 - 2 | 218 | 69 | |
XM_015784464.2 | XP_015639950.1 | 1 - 2 | 218 | 69 | ||
GenBank mRNA | AK070757.1 | - | 1 - 2 | 171 | 69 | |
AK109378.2 | - | 1 - 2 | 199 | 69 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015784463.2, XM_015784464.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
