Assay Details
Gene Symbol: LOC4338690 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015785053.2 | XP_015640539.1 | 3 - 4 | 4713 | 67 | |
XM_015785054.2 | XP_015640540.1 | 4 - 5 | 4622 | 67 | ||
XM_015785056.2 | XP_015640542.1 | 4 - 5 | 4556 | 67 | ||
XM_015785057.2 | XP_015640543.1 | 4 - 5 | 4571 | 67 | ||
GenBank mRNA | AK068327.1 | - | 2 - 3 | 1490 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
