Assay Details
Gene Symbol: LOC4338944 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015783899.2 | XP_015639385.1 | 3 - 4 | 2901 | 151 | |
XM_015783900.2 | XP_015639386.1 | 3 - 4 | 2898 | 151 | ||
XM_015783901.2 | XP_015639387.1 | 3 - 4 | 2905 | 151 | ||
XM_015783902.2 | XP_015639388.1 | 3 - 4 | 2905 | 151 |
Gene Symbol: LOC4338940 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015784881.2 | XP_015640367.1 | 3 - 4 | 2935 | 151 | |
XM_015784882.2 | XP_015640368.1 | 3 - 4 | 2932 | 151 | ||
XM_015784883.2 | XP_015640369.1 | 3 - 4 | 2941 | 151 | ||
XM_015784884.2 | XP_015640370.1 | 3 - 4 | 2944 | 151 | ||
GenBank mRNA | AK070640.1 | - | 3 - 4 | 2886 | 151 | |
FJ536320.1 | - | 2 - 3 | 2572 | 151 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
