Assay Details
Gene Symbol: LOC4339216 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015783880.2 | XP_015639366.1 | 1 - 2 | 159 | 115 | |
GenBank mRNA | AK058219.1 | - | 1 - 2 | 93 | 115 | |
BT128929.1 | - | 1 - 2 | 92 | 115 | ||
BT129141.1 | - | 1 - 2 | 92 | 115 | ||
BT129342.1 | - | 1 - 2 | 89 | 115 | ||
BT129822.1 | - | 1 - 2 | 86 | 115 | ||
BT130575.1 | - | 1 - 2 | 86 | 115 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015783880.2 transcripts.
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
