Assay Details
| Gene Symbol: LOC4341032 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015788115.1 | XP_015643601.1 | 2 - 3 | 484 | 71 | |
| GenBank mRNA | AK099499.1 | - | 2 - 3 | 442 | 71 | |
| AK111525.1 | - | 2 - 3 | 433 | 71 | ||
More Information
Set Membership: |
|

LOC4341032
Genomic Map