Assay Details
Gene Symbol: LOC4341186 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015788704.2 | XP_015644190.1 | 5 - 6 | 2007 | 109 | |
GenBank mRNA | AK241349.1 | - | 1 - 2 | 63 | 109 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
