Assay Details
Gene Symbol: LOC4341186 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015788704.2 | XP_015644190.1 | 6 - 7 | 2093 | 74 | |
GenBank mRNA | AK241349.1 | - | 2 - 3 | 149 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
