Assay Details
| Gene Symbol: LOC4341197 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015788961.1 | XP_015644447.1 | 3 - 4 | 668 | 109 | |
| GenBank mRNA | AK071835.1 | - | 3 - 4 | 675 | 109 | |
More Information
Set Membership: |
|

LOC4341197
Genomic Map