Assay Details
| Gene Symbol: LOC4337632 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015782702.2 | XP_015638188.1 | 1 | 948 | 108 | |
| GenBank mRNA | AK110694.1 | - | 1 | 954 | 108 | |
More Information
Set Membership: |
|

LOC4337632
Genomic Map