Assay Details
| Gene Symbol: LOC4339642 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015783983.2 | XP_015639469.1 | 1 - 2 | 916 | 70 | |
| XM_015783984.2 | XP_015639470.1 | 1 - 2 | 916 | 70 | ||
| GenBank mRNA | AK121133.1 | - | 1 - 2 | 912 | 70 | |
More Information
Set Membership: |
|

LOC4339642
Genomic Map