Assay Details
| Gene Symbol: LOC4339642 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015783984.2 | XP_015639470.1 | 4 - 5 | 2195 | 133 | |
| GenBank mRNA | AK121133.1 | - | 4 - 5 | 2191 | 133 | |
More Information
Set Membership: |
|

LOC4339642
Genomic Map