Assay Details
| Gene Symbol: LOC4341464 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015785567.2 | XP_015641053.1 | 3 - 4 | 1659 | 76 | |
| XM_015785570.2 | XP_015641056.1 | 5 - 6 | 1462 | 76 | ||
| XM_015785573.2 | XP_015641059.1 | 5 - 6 | 1474 | 76 | ||
| XM_015785574.2 | XP_015641060.1 | 4 - 5 | 1234 | 76 | ||
| GenBank mRNA | AK106905.1 | - | 4 - 5 | 1284 | 76 | |
More Information
Set Membership: |
|

LOC4341464
Genomic Map