Assay Details
Gene Symbol: LOC4341477 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015787598.1 | XP_015643084.1 | 11 - 12 | 1230 | 75 | |
GenBank mRNA | AK102270.1 | - | 11 - 12 | 1230 | 75 | |
EU703803.1 | - | 11 - 12 | 1131 | 75 | ||
EU703804.1 | - | 10 - 11 | 1086 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
