Assay Details
Gene Symbol: LOC4341477 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015787598.1 | XP_015643084.1 | 1 - 2 | 285 | 120 | |
GenBank mRNA | AK102270.1 | - | 1 - 2 | 285 | 120 | |
EU703803.1 | - | 1 - 2 | 186 | 120 | ||
EU703804.1 | - | 141 | 120 |

More Information
Set Membership: |
![]() ![]() ![]() |
