Assay Details
Gene Symbol: LOC4341477 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015787598.1 | XP_015643084.1 | 2 - 3 | 349 | 129 | |
GenBank mRNA | AK102270.1 | - | 2 - 3 | 349 | 129 | |
EU703803.1 | - | 2 - 3 | 250 | 129 | ||
EU703804.1 | - | 1 - 2 | 205 | 129 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
