Assay Details
Gene Symbol: LOC4341477 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015787598.1 | XP_015643084.1 | 5 - 6 | 600 | 114 | |
GenBank mRNA | AK102270.1 | - | 5 - 6 | 600 | 114 | |
EU703803.1 | - | 5 - 6 | 501 | 114 | ||
EU703804.1 | - | 4 - 5 | 456 | 114 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
