Assay Details
Gene Symbol: LOC4341477 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015787598.1 | XP_015643084.1 | 8 - 9 | 874 | 122 | |
GenBank mRNA | AK102270.1 | - | 8 - 9 | 874 | 122 | |
EU703803.1 | - | 8 - 9 | 775 | 122 | ||
EU703804.1 | - | 7 - 8 | 730 | 122 |

More Information
Set Membership: |
![]() ![]() ![]() |
