Assay Details
Gene Symbol: LOC4341477 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015787598.1 | XP_015643084.1 | 9 - 10 | 994 | 79 | |
GenBank mRNA | AK102270.1 | - | 9 - 10 | 994 | 79 | |
EU703803.1 | - | 9 - 10 | 895 | 79 | ||
EU703804.1 | - | 8 - 9 | 850 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
