Assay Details
| Gene Symbol: LOC4341687 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015788012.2 | XP_015643498.1 | 1 - 2 | 715 | 116 | |
| GenBank mRNA | AK071905.1 | - | 1 - 2 | 665 | 116 | |
| AK099071.1 | - | 1 - 2 | 593 | 116 | ||
| AK099209.1 | - | 1 - 2 | 665 | 116 | ||
More Information
Set Membership: |
|

LOC4341687
Genomic Map