Assay Details
Gene Symbol: LOC4342175 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015789084.2 | XP_015644570.1 | 2 - 3 | 380 | 75 | |
GenBank mRNA | AK101907.1 | - | 2 - 3 | 381 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
